Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015378.3 | NP_056193.2 | 31 | 7324 | ||
NM_018156.3 | NP_060626.2 | 31 | 7324 | |||
GenBank mRNA | AB007922.3 | - | 13 | 3634 | ||
AJ608774.1 | - | 31 | 7296 | |||
AJ608775.1 | - | 31 | 7296 | |||
AK001481.1 | - | 4 | 444 | |||
AK125118.1 | - | 7 | 1054 | |||
BX641035.1 | - | 6 | 888 |