Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111018.1 | NP_001104488.1 | 2 | 371 | ||
NM_001244963.1 | NP_001231892.1 | 2 | 695 | |||
NM_145117.4 | NP_660093.2 | 2 | 695 | |||
NM_182964.5 | NP_892009.3 | 2 | 695 | |||
XM_006718364.3 | XP_006718427.1 | 2 | 1017 | |||
XM_006718365.3 | XP_006718428.1 | 2 | 1017 | |||
XM_006718366.3 | XP_006718429.1 | 2 | 181 | |||
XM_011520444.2 | XP_011518746.1 | 2 | 1017 | |||
XM_011520445.2 | XP_011518747.1 | 2 | 1017 | |||
XM_011520446.2 | XP_011518748.1 | 2 | 1017 | |||
XM_011520447.2 | XP_011518749.1 | 2 | 1017 | |||
XM_011520448.2 | XP_011518750.1 | 2 | 888 | |||
XM_011520449.2 | XP_011518751.1 | 2 | 1017 | |||
XM_011520450.2 | XP_011518752.1 | 2 | 1017 | |||
XM_011520451.2 | XP_011518753.1 | 2 | 464 | |||
XM_011520452.2 | XP_011518754.1 | 3 | 834 | |||
XM_017018520.1 | XP_016874009.1 | 3 | 809 | |||
XM_017018521.1 | XP_016874010.1 | 2 | 1017 | |||
XM_017018522.1 | XP_016874011.1 | 3 | 834 | |||
XM_017018523.1 | XP_016874012.1 | 2 | 1694 | |||
XM_017018524.1 | XP_016874013.1 | 2 | 517 | |||
GenBank mRNA | AB037840.3 | - | 2 | 696 | ||
AF466143.1 | - | 2 | 410 | |||
AF466144.1 | - | 2 | 410 | |||
BC150290.1 | - | 2 | 696 | 8860130 |