Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 5 | 614 | ||
XM_017000413.1 | XP_016855902.1 | 4 | 763 | |||
XM_017000414.1 | XP_016855903.1 | 3 | 605 | |||
GenBank mRNA | BC041458.1 | - | 3 | 1132 | ||
BC128073.1 | - | 5 | 614 | 40109068 |