Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26453
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
7
736
NM_001321198.1
NP_001308127.1
7
617
NM_001321199.1
NP_001308128.1
7
581
NM_001321200.1
NP_001308129.1
7
637
NM_001321201.1
NP_001308130.1
6
509
NM_018224.3
NP_060694.2
6
516
NR_047701.1
7
581
NR_135580.1
7
617
NR_135581.1
7
617
NR_135582.1
6
516
NR_135583.1
6
516
XM_005249807.4
XP_005249864.1
6
580
XM_006715752.2
XP_006715815.1
7
548
XM_017012414.1
XP_016867903.1
6
507
XM_017012415.1
XP_016867904.1
7
609
XM_017012416.1
XP_016867905.1
6
507
XM_017012417.1
XP_016867906.1
6
507
XM_017012418.1
XP_016867907.1
6
507
XM_017012419.1
XP_016867908.1
6
507
XM_017012421.1
XP_016867910.1
7
731
XM_017012422.1
XP_016867911.1
7
620
XM_017012423.1
XP_016867912.1
7
540
XM_017012424.1
XP_016867913.1
6
523
XM_017012425.1
XP_016867914.1
7
624
XM_017012426.1
XP_016867915.1
7
743
XM_017012427.1
XP_016867916.1
8
698
XM_017012428.1
XP_016867917.1
6
523
XM_017012429.1
XP_016867918.1
5
417
XM_017012430.1
XP_016867919.1
7
609
XM_017012431.1
XP_016867920.1
6
523
XM_017012432.1
XP_016867921.1
6
523
GenBank mRNA
AK001665.1
-
7
550
AK021905.1
-
7
686
AK023007.1
-
7
533
AK023053.1
-
7
549
AK097203.1
-
5
2047
AK129690.1
-
3
1240
AK293043.1
-
7
559
BC056884.1
-
7
508
6159045
BG028456.1
-
538
BG334368.1
-
7
526
BG548154.1
-
7
573
BM918741.1
-
6
489
BQ231567.1
-
6
454
DA742258.1
-
6
498
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0