Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZBTB7C SIRNA Assays ›
Gene Symbol:
ZBTB7C
Gene Name:
zinc finger and BTB domain containing 7C
Gene Aliases:
APM-1, APM1, ZBTB36, ZNF857C
Chromosome Location:
Chr.18: 48026886 - 48412639 on Build GRCh38
UniGene ID:
Hs.515388
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
264587
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZBTB7C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039360.2
NP_001034449.1
2
348
NM_001318841.1
NP_001305770.1
4
529
XM_005258229.4
XP_005258286.1
3
273
XM_011525861.2
XP_011524163.1
6
753
XM_011525863.2
XP_011524165.1
5
870
XM_011525864.2
XP_011524166.1
5
625
XM_011525865.2
XP_011524167.1
5
866
XM_011525866.2
XP_011524168.1
4
611
XM_011525867.2
XP_011524169.1
4
400
XM_011525869.2
XP_011524171.1
5
875
XM_011525870.2
XP_011524172.1
4
571
XM_011525871.2
XP_011524173.1
2
1086
XM_017025605.1
XP_016881094.1
6
2548
XM_017025606.1
XP_016881095.1
6
1002
XM_017025607.1
XP_016881096.1
6
757
XM_017025608.1
XP_016881097.1
5
2425
XM_017025609.1
XP_016881098.1
4
742
GenBank mRNA
AK296709.1
-
2
348
AM988836.1
-
2
336
AM988837.1
-
2
481
AM988838.1
-
3
531
AM988839.1
-
4
529
AM989328.1
-
5
538
AM989329.1
-
4
403
AM989330.1
-
5
547
AM989331.1
-
6
558
AM989332.1
-
2
499
AM991276.1
-
4
399
AM991277.1
-
5
526
AM991278.1
-
5
550
AM991279.1
-
6
639
AM991280.1
-
6
679
AM991281.1
-
6
658
AM991282.1
-
3
174
AM991283.1
-
4
301
AM991284.1
-
5
351
AM991285.1
-
4
472
AM991959.1
-
4
359
AM991960.1
-
5
486
AM991961.1
-
5
482
AM991962.1
-
6
536
AM991963.1
-
6
532
AM991964.1
-
5
491
AM991965.1
-
6
550
AM991966.1
-
6
614
AM991967.1
-
5
451
AM991968.1
-
6
547
AM991969.1
-
6
549
AM991970.1
-
9
875
AM998476.1
-
325
AM998477.1
-
4
416
AM998478.1
-
4
448
AM998479.1
-
4
452
AM998480.1
-
4
457
AM998481.1
-
6
524
AM998482.1
-
5
520
AM998483.1
-
5
554
AM998484.1
-
5
584
AM998485.1
-
5
580
AM998486.1
-
5
605
AM998487.1
-
6
634
AM998488.1
-
2
246
AM998489.1
-
3
375
EF120357.1
-
2
400
EU669438.1
-
4
382
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
regulation of RNA polymerase II regulatory region sequence-specific DNA binding
nucleic acid binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0