Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 18 | 3745 | ||
XM_006718795.3 | XP_006718858.1 | 7 | 2259 | |||
XM_011542691.2 | XP_011540993.1 | 19 | 4432 | |||
XM_011542692.2 | XP_011540994.1 | 19 | 4420 | |||
XM_011542694.2 | XP_011540996.1 | 19 | 4435 | |||
XM_011542695.2 | XP_011540997.1 | 18 | 3963 | |||
XM_017017393.1 | XP_016872882.1 | 18 | 4450 | |||
XM_017017394.1 | XP_016872883.1 | 19 | 4435 | |||
XM_017017395.1 | XP_016872884.1 | 19 | 4434 | |||
XM_017017396.1 | XP_016872885.1 | 12 | 3774 | |||
GenBank mRNA | AB028953.1 | - | 6 | 1987 |