Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 23 | 4445 | ||
XM_005264722.1 | XP_005264779.1 | 23 | 4122 | |||
XM_005264723.1 | XP_005264780.1 | 13 | 3604 | |||
XM_017005989.1 | XP_016861478.1 | 24 | 4321 | |||
XM_017005990.1 | XP_016861479.1 | 24 | 4214 | |||
XM_017005991.1 | XP_016861480.1 | 25 | 4374 | |||
XM_017005992.1 | XP_016861481.1 | 24 | 4154 | |||
XM_017005993.1 | XP_016861482.1 | 23 | 4053 | |||
XM_017005994.1 | XP_016861483.1 | 23 | 5895 | |||
XM_017005995.1 | XP_016861484.1 | 18 | 3731 | |||
XM_017005996.1 | XP_016861485.1 | 14 | 3433 | |||
XM_017005997.1 | XP_016861486.1 | 14 | 3696 | |||
GenBank mRNA | AB023230.1 | - | 23 | 4214 | ||
AK024086.1 | - | 7 | 2480 | |||
AL110231.1 | - | 1 | 488 | |||
AL832231.1 | - | 13 | 3442 | |||
BC028291.1 | - | 3 | 1794 | |||
BC110637.1 | - | 8 | 2666 |