Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015175.2 | NP_055990.1 | 20 | 3073 | ||
XM_006713072.3 | XP_006713135.1 | 19 | 2989 | |||
XM_011533533.2 | XP_011531835.1 | 20 | 3070 | |||
XM_017006010.1 | XP_016861499.1 | 20 | 3070 | |||
XM_017006011.1 | XP_016861500.1 | 20 | 2926 | |||
XM_017006012.1 | XP_016861501.1 | 19 | 2989 | |||
XM_017006013.1 | XP_016861502.1 | 20 | 3070 | |||
XM_017006014.1 | XP_016861503.1 | 19 | 2845 | |||
XM_017006015.1 | XP_016861504.1 | 18 | 2701 | |||
XM_017006016.1 | XP_016861505.1 | 15 | 3647 | |||
GenBank mRNA | AB011112.3 | - | 8 | 1309 | ||
AK131104.1 | - | 20 | 2913 |