Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 12 | 4136 | ||
XM_011538078.2 | XP_011536380.1 | 13 | 4138 | |||
XM_017019083.1 | XP_016874572.1 | 12 | 4006 | |||
XM_017019084.1 | XP_016874573.1 | 14 | 3797 | |||
XM_017019085.1 | XP_016874574.1 | 14 | 4017 | |||
XM_017019086.1 | XP_016874575.1 | 14 | 4022 | |||
XM_017019087.1 | XP_016874576.1 | 12 | 3888 | |||
GenBank mRNA | AB023201.1 | - | 13 | 3730 |