Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018130.2 | NP_060600.2 | 11 | 1408 | ||
GenBank mRNA | AK001401.1 | - | 11 | 1406 | ||
AK092962.1 | - | 11 | 1406 | |||
AK296789.1 | - | 11 | 1307 | |||
BC017204.2 | - | 11 | 1398 | 3617948 | ||
BC017274.2 | - | 11 | 1386 | 4840343 | ||
BC025270.1 | - | 11 | 1398 | 4622871 | ||
BC032671.1 | - | 11 | 1412 | 5538687 | ||
BC039830.1 | - | 11 | 1341 | 6064610 | ||
BC047879.2 | - | 11 | 1359 | 5541019 | ||
BX641074.1 | - | 3 | 2583 |