Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPIDR SIRNA Assays ›
Gene Symbol:
SPIDR
Gene Name:
scaffolding protein involved in DNA repair
Gene Aliases:
KIAA0146
Chromosome Location:
Chr.8: 47260878 - 47736001 on Build GRCh38
UniGene ID:
Hs.381058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
264796
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPIDR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080394.3
NP_001073863.1
20
2849
NM_001282916.1
NP_001269845.1
19
2782
NM_001282919.1
NP_001269848.1
19
2813
NR_104581.1
17
2246
XM_005251189.3
XP_005251246.1
20
2846
XM_005251191.3
XP_005251248.1
17
2528
XM_005251193.3
XP_005251250.1
18
2622
XM_005251195.4
XP_005251252.1
19
2635
XM_005251198.4
XP_005251255.1
19
2569
XM_005251199.4
XP_005251256.1
17
2383
XM_006716443.3
XP_006716506.1
20
2759
XM_006716444.3
XP_006716507.1
19
2532
XM_011517497.2
XP_011515799.1
21
2922
XM_011517500.2
XP_011515802.1
20
2689
XM_011517507.2
XP_011515809.1
12
1380
XM_017013268.1
XP_016868757.1
19
2792
XM_017013269.1
XP_016868758.1
18
2732
XM_017013270.1
XP_016868759.1
19
2690
XM_017013271.1
XP_016868760.1
19
2907
XM_017013272.1
XP_016868761.1
15
2536
XM_017013273.1
XP_016868762.1
17
2350
XM_017013274.1
XP_016868763.1
18
2636
XM_017013275.1
XP_016868764.1
15
2388
XM_017013276.1
XP_016868765.1
15
2375
XM_017013277.1
XP_016868766.1
18
2607
XM_017013278.1
XP_016868767.1
19
2601
XM_017013279.1
XP_016868768.1
19
2780
XM_017013280.1
XP_016868769.1
18
2570
XM_017013281.1
XP_016868770.1
18
2535
XM_017013282.1
XP_016868771.1
16
2372
XM_017013283.1
XP_016868772.1
20
2834
XM_017013284.1
XP_016868773.1
17
2466
XM_017013285.1
XP_016868774.1
19
2624
XM_017013286.1
XP_016868775.1
16
2384
XM_017013287.1
XP_016868776.1
17
2402
XM_017013288.1
XP_016868777.1
15
2153
XM_017013289.1
XP_016868778.1
16
2344
XM_017013290.1
XP_016868779.1
15
2214
XM_017013291.1
XP_016868780.1
21
2814
GenBank mRNA
AK026929.1
-
11
1644
AK055680.1
-
13
1411
AK092336.1
-
1
2031
AK122820.1
-
21
3142
AK294275.1
-
19
2704
AK301677.1
-
11
1286
AK303140.1
-
10
1491
AK303579.1
-
19
2760
BC015561.2
-
6
669
BC031627.1
-
17
2171
D63480.1
-
20
2777
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
positive regulation of protein complex assembly
regulation of establishment of protein localization to chromosome
cellular response to ionizing radiation
cellular response to hydroxyurea
cellular response to camptothecin
positive regulation of double-strand break repair
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0