Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077657.2 | NP_001071125.1 | 3 | 1490 | ||
NM_001320473.1 | NP_001307402.1 | 3 | 1546 | |||
GenBank mRNA | AK123917.1 | - | 3 | 1490 | ||
AL833140.1 | - | 3 | 1471 | |||
AL833443.1 | - | 3 | 1526 |