Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018149.6 | NP_060619.4 | 4 | 2963 | ||
GenBank mRNA | AF349467.1 | - | 4 | 3059 | ||
AK001449.1 | - | 4 | 2112 | |||
AK026858.1 | - | 4 | 2936 | |||
AL834490.1 | - | 4 | 2942 | |||
BC020957.1 | - | 4 | 2131 | 3851898 |