Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 10 | 2691 | ||
XM_006716754.3 | XP_006716817.1 | 8 | 1246 | |||
XM_017014618.1 | XP_016870107.1 | 10 | 2897 | |||
XM_017014619.1 | XP_016870108.1 | 10 | 3008 | |||
GenBank mRNA | AB209610.1 | - | 9 | 2279 | ||
AK123986.1 | - | 2 | 365 | |||
BC125124.1 | - | 8 | 1285 | 40119018 | ||
BC125125.1 | - | 10 | 2770 | 40119019 | ||
BC140785.1 | - | 9 | 2662 | 9021653 | ||
BC171785.1 | - | 9 | 2662 | 9054439 |