Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015352.1 | 2 | 884 | |||
GenBank mRNA | AK098045.1 | - | 2 | 215 | ||
AY026052.1 | - | 2 | 884 | |||
BC043198.2 | - | 2 | 170 |