Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251904.1 | NP_001238833.1 | 12, 13 | 1327 | ||
NM_001251905.1 | NP_001238834.1 | 12, 13 | 1352 | |||
NM_018171.3 | NP_060641.2 | 12, 13 | 1309 | |||
XM_006719472.1 | XP_006719535.1 | 12, 13 | 1246 | |||
XM_011538530.2 | XP_011536832.1 | 12, 13 | 2779 | |||
XM_011538531.2 | XP_011536833.1 | 12, 13 | 2442 | |||
XM_011538532.2 | XP_011536834.1 | 12, 13 | 3768 | |||
XM_017019551.1 | XP_016875040.1 | 12, 13 | 1714 | |||
XM_017019552.1 | XP_016875041.1 | 12, 13 | 2424 | |||
XM_017019553.1 | XP_016875042.1 | 13, 14 | 4022 | |||
XM_017019554.1 | XP_016875043.1 | 12, 13 | 1228 | |||
GenBank mRNA | AK001521.2 | - | 12, 13 | 1129 | ||
AK290562.1 | - | 12, 13 | 1162 | |||
AK293783.1 | - | 12, 13 | 1101 | |||
AK296610.1 | - | 12, 13 | 1108 | |||
AK297100.1 | - | 12, 13 | 1180 | |||
AK310377.1 | - | 12, 13 | 1097 | |||
AY113704.1 | - | 12, 13 | 1309 | |||
BC033731.1 | - | 12, 13 | 1213 | 5168590 |