Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085423.1 | NP_001078892.1 | 4 | 551 | ||
NM_001291317.1 | NP_001278246.1 | 3 | 281 | |||
XM_017024486.1 | XP_016879975.1 | 4 | 644 | |||
XM_017024487.1 | XP_016879976.1 | 4 | 564 | |||
GenBank mRNA | AB542950.1 | - | 4 | 503 | ||
AB542952.1 | - | 3 | 227 | |||
BC018972.1 | - | 4 | 536 | |||
BC053534.1 | - | 3 | 445 | |||
BC130406.1 | - | 4 | 526 | 40146437 | ||
BC136594.1 | - | 4 | 526 | 9020584 | ||
CD687595.1 | - | 4 | 566 |