Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018178.5 | NP_060648.2 | 5 | 1560 | ||
XM_006711428.1 | XP_006711491.1 | 5 | 1487 | |||
GenBank mRNA | AJ296153.1 | - | 5 | 1388 | ||
AK001549.1 | - | 4 | 1188 | |||
BC013870.2 | - | 4 | 1359 | 3837466 |