Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014283.1 | NP_001014305.1 | 7 | 843 | ||
XM_017020643.1 | XP_016876132.1 | 7 | 1054 | |||
XM_017020644.1 | XP_016876133.1 | 7 | 984 | |||
XM_017020645.1 | XP_016876134.1 | 7 | 1025 | |||
XM_017020646.1 | XP_016876135.1 | 7 | 4766 | |||
XM_017020647.1 | XP_016876136.1 | 7 | 1304 | |||
XM_017020648.1 | XP_016876137.1 | 7 | 2067 | |||
XM_017020649.1 | XP_016876138.1 | 6 | 881 | |||
GenBank mRNA | AK000099.1 | - | 5 | 440 | ||
AK001566.1 | - | 8 | 978 | |||
BC056669.1 | - | 7 | 830 | 6494926 |