Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 9 | 1844 | ||
NM_018212.4 | NP_060682.2 | 9 | 1844 | |||
XM_005273187.2 | XP_005273244.1 | 10 | 1884 | |||
XM_011544229.2 | XP_011542531.2 | 10 | 2360 | |||
XM_017001746.1 | XP_016857235.1 | 11 | 3119 | |||
XM_017001747.1 | XP_016857236.1 | 11 | 2372 | |||
XM_017001748.1 | XP_016857237.1 | 9 | 2303 | |||
XM_017001749.1 | XP_016857238.1 | 10 | 2360 | |||
XM_017001750.1 | XP_016857239.1 | 10 | 2261 | |||
XM_017001751.1 | XP_016857240.1 | 9 | 2192 | |||
XM_017001752.1 | XP_016857241.1 | 8 | 2192 | |||
XM_017001753.1 | XP_016857242.1 | 12 | 1923 | |||
XM_017001754.1 | XP_016857243.1 | 11 | 1867 | |||
XM_017001755.1 | XP_016857244.1 | 11 | 1610 | |||
XM_017001756.1 | XP_016857245.1 | 9 | 1541 | |||
XM_017001757.1 | XP_016857246.1 | 9 | 1444 | |||
GenBank mRNA | AF519769.1 | - | 9 | 1660 | ||
AK001635.1 | - | 3 | 324 | |||
AK096246.1 | - | 7 | 1140 | |||
AK126894.1 | - | 4 | 601 | |||
AK127561.1 | - | 3 | 975 | |||
AK290179.1 | - | 9 | 1788 | |||
AL133059.1 | - | 3 | 215 | |||
AY345143.1 | - | 9 | 1748 | |||
BC065238.1 | - | 8 | 1262 | |||
BC095481.1 | - | 9 | 1843 | 30348815 | ||
EU255274.1 | - | 7 | 1284 | |||
KM214203.1 | - | 9 | 1448 | |||
KM214204.1 | - | 9 | 1448 |