Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145531.2 | NP_001139003.1 | 4 | 1265 | ||
NM_001302625.1 | NP_001289554.1 | 3 | 605 | |||
XM_017012010.1 | XP_016867499.1 | 4 | 3755 | |||
XM_017012011.1 | XP_016867500.1 | 5 | 3641 | |||
XM_017012012.1 | XP_016867501.1 | 4 | 3367 | |||
XM_017012013.1 | XP_016867502.1 | 5 | 3283 | |||
XM_017012014.1 | XP_016867503.1 | 4 | 3363 | |||
GenBank mRNA | AK093321.1 | - | 4 | 1265 |