Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104925.1 | NP_001098395.1 | 6 | 3562 | ||
NM_018257.2 | NP_060727.2 | 6 | 3643 | |||
GenBank mRNA | AK001745.1 | - | 6 | 3572 | ||
AK074436.1 | - | 1 | 2660 | |||
BC033665.2 | - | 5 | 3468 | 4999114 |