Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 2 | 448 | ||
XM_005248551.4 | XP_005248608.1 | 2 | 415 | |||
XM_005248552.3 | XP_005248609.1 | 2 | 415 | |||
XM_011543505.2 | XP_011541807.1 | 2 | 210 | |||
XM_011543507.2 | XP_011541809.1 | 2 | 415 | |||
XM_017009606.1 | XP_016865095.1 | 2 | 858 | |||
GenBank mRNA | AF115511.1 | - | 2 | 197 | ||
AK001766.1 | - | 2 | 394 | |||
AK021855.1 | - | 2 | 234 | |||
AK023977.1 | - | 2 | 150 | |||
AK293316.1 | - | 2 | 107 | |||
AK295249.1 | - | 2 | 128 | |||
AK296423.1 | - | 2 | 224 | |||
AK304329.1 | - | 2 | 130 | |||
BC040241.1 | - | 2 | 200 | 3447269 | ||
BM818066.1 | - | 2 | 415 | |||
DC403958.1 | - | 2 | 449 | |||
KU178690.1 | - | 2 | 128 | |||
KU178691.1 | - | 2 | 128 |