Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303470.1 | NP_001290399.1 | 2, 3 | 231 | ||
NM_018275.4 | NP_060745.3 | 2, 3 | 731 | |||
GenBank mRNA | AK054872.1 | - | 2, 3 | 227 | ||
AK310170.1 | - | 3, 4 | 495 | |||
BC002749.2 | - | 2, 3 | 131 | |||
BC015722.2 | - | 2, 3 | 689 | 3920412 |