Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018279.3 | NP_060749.2 | 2, 3 | 835 | ||
GenBank mRNA | AK001798.1 | - | 2, 3 | 437 | ||
AK223148.1 | - | 2, 3 | 433 | |||
AK315587.1 | - | 2, 3 | 394 | |||
BC008596.1 | - | 2, 3 | 835 | 4154303 |