siRNA Details
Gene Symbol: ECHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198961.1 | NP_001185890.1 | 1, 2 | 230 | ||
NM_001198962.1 | NP_001185891.1 | 1, 2 | 230 | |||
NM_018281.3 | NP_060751.2 | 1, 2 | 230 | |||
XM_011541709.2 | XP_011540011.1 | 2, 3 | 441 | |||
XM_011541710.2 | XP_011540012.1 | 2, 3 | 441 | |||
XM_011541711.2 | XP_011540013.1 | 2, 3 | 441 | |||
XM_011541712.2 | XP_011540014.1 | 2, 3 | 441 | |||
XM_011541720.2 | XP_011540022.1 | 1, 2 | 3987 | |||
XM_011541721.2 | XP_011540023.1 | 3, 4 | 680 | |||
XM_011541722.2 | XP_011540024.1 | 2, 3 | 298 | |||
XM_011541723.2 | XP_011540025.1 | 2, 3 | 398 | |||
XM_011541724.2 | XP_011540026.1 | 2, 3 | 441 | |||
XM_011541725.2 | XP_011540027.1 | 2, 3 | 441 | |||
XM_011541727.2 | XP_011540029.1 | 1, 2 | 3987 | |||
XM_017001641.1 | XP_016857130.1 | 3, 4 | 1821 | |||
GenBank mRNA | AK001810.1 | - | 1, 2 | 182 | ||
AK299834.1 | - | 2, 3 | 399 | |||
AK300896.1 | - | 2, 3 | 269 | |||
AK302285.1 | - | 4, 5 | 809 | |||
AK307381.1 | - | 1, 2 | 125 | |||
AK310650.1 | - | 2, 3 | 362 | |||
AK315906.1 | - | 2, 3 | 361 | |||
BC044574.1 | - | 1, 2 | 160 | 5314083 | ||
BC051887.1 | - | 1, 2 | 218 | |||
CF127686.1 | - | 1, 2 | 174 |

More Information
Panther Classification:
Molecular Function -










