Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145020.1 | NP_001138492.1 | 5 | 474 | ||
NR_026736.1 | 6 | 539 | ||||
NR_026737.1 | 4 | 392 | ||||
XM_006715088.3 | XP_006715151.1 | 6 | 733 | |||
XM_011514572.2 | XP_011512874.1 | 5 | 529 | |||
XM_011514574.2 | XP_011512876.1 | 4 | 439 | |||
XM_011514575.2 | XP_011512877.1 | 6 | 715 | |||
GenBank mRNA | BC016820.1 | - | 4 | 288 |