Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282275.1 | NP_001269204.1 | 2 | 358 | ||
NM_001282276.1 | NP_001269205.1 | 2 | 292 | |||
XM_006721865.3 | XP_006721928.1 | 2 | 974 | |||
XM_011524747.2 | XP_011523049.1 | 2 | 284 | |||
XM_011524748.1 | XP_011523050.1 | 2 | 229 | |||
GenBank mRNA | AF532777.1 | - | 2 | 346 | ||
BX106051.1 | - | 2 | 292 |
Gene Symbol: LOC102724665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011600.1 | XP_016867089.1 | 1 | 498 |