Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099667.1 | NP_001093137.1 | 2 | 389 | ||
GenBank mRNA | BC066349.1 | - | 2 | 389 | ||
BC090924.1 | - | 2 | 387 |