siRNA Details
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 25 | 9129 | ||
GenBank mRNA | AB290171.1 | - | 12 | 5829 | ||
BX647806.1 | - | 5 | 1806 |

Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 25 | 9129 | ||
GenBank mRNA | AB290171.1 | - | 12 | 5829 | ||
BX647806.1 | - | 5 | 1806 |