Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145026.1 | NP_001138498.1 | 9 | 1266 | ||
XM_017019273.1 | XP_016874762.1 | 13 | 2068 | |||
XM_017019274.1 | XP_016874763.1 | 13 | 2069 | |||
XM_017019275.1 | XP_016874764.1 | 13 | 2069 | |||
GenBank mRNA | DV080649.1 | - | 2 | 217 |
Set Membership: |
G-protein coupled receptor |