Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002726.2 | 1 | 1337 |
Gene Symbol: HNRNPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194247.2 | NP_919223.1 | 11 | 1411 | ||
XM_005246381.4 | XP_005246438.1 | 11 | 1317 | |||
XM_006712365.2 | XP_006712428.1 | 10 | 1270 | |||
GenBank mRNA | AF517524.1 | - | 4 | 360 | ||
AK293125.1 | - | 11 | 1279 | |||
AK303208.1 | - | 6 | 617 | |||
AK304687.1 | - | 11 | 1175 | |||
AK316434.1 | - | 11 | 1279 | |||
BC064494.1 | - | 9 | 1026 | |||
BC080591.1 | - | 7 | 739 |