Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099455.1 | NP_001092925.1 | 2 | 194 | ||
NM_018340.2 | NP_060810.2 | 2 | 194 | |||
GenBank mRNA | AK002013.1 | - | 2 | 177 | ||
AK022710.1 | - | 2 | 144 | |||
AK093854.1 | - | 2 | 190 | |||
AK225359.1 | - | 2 | 144 | |||
AK225617.1 | - | 2 | 189 | |||
AK298662.1 | - | 2 | 119 | |||
AY553877.1 | - | 2 | 83 | |||
BC006289.2 | - | 2 | 178 | 3959701 | ||
BX640805.1 | - | 2 | 160 | |||
DA387854.1 | - | 2 | 194 | |||
DB468489.1 | - | 2 | 136 |