Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039614.2 | NP_001034703.1 | 2 | 1203 | ||
NM_001303254.1 | NP_001290183.1 | 3 | 1086 | |||
XM_005254369.4 | XP_005254426.1 | 3 | 2486 | |||
GenBank mRNA | BC111368.1 | - | 2 | 1203 | 7504115 | |
CK823338.1 | - | 1 | 522 |