Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310124.1 | NP_001297053.1 | 13 | 2059 | ||
XM_011527571.2 | XP_011525873.1 | 13 | 1671 | |||
XM_011527573.2 | XP_011525875.1 | 12 | 1839 | |||
XM_011527574.2 | XP_011525876.1 | 12 | 1566 |