Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080418.2 | NP_001073887.1 | 2 | 160 | ||
XM_011541879.2 | XP_011540181.1 | 3 | 2045 |