Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080473.1 | NP_001073942.1 | 2 | 154 | ||
XM_005264301.4 | XP_005264358.1 | 2 | 1215 | |||
XM_017004063.1 | XP_016859552.1 | 2 | 1215 | |||
XM_017004064.1 | XP_016859553.1 | 2 | 1215 | |||
XM_017004065.1 | XP_016859554.1 | 2 | 1215 | |||
XM_017004066.1 | XP_016859555.1 | 2 | 1215 | |||
XM_017004067.1 | XP_016859556.1 | 2 | 1215 | |||
XM_017004068.1 | XP_016859557.1 | 2 | 1215 | |||
XM_017004069.1 | XP_016859558.1 | 2 | 1215 | |||
XM_017004070.1 | XP_016859559.1 | 2 | 1215 | |||
GenBank mRNA | BC033385.1 | - | 2 | 195 |