Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128591.1 | NP_001122063.1 | 3 | 704 | ||
NM_001128592.1 | NP_001122064.1 | 4 | 821 | |||
XM_011514591.2 | XP_011512893.1 | 4 | 1256 | |||
GenBank mRNA | AK126623.1 | - | 2 | 4495 | ||
BX648600.1 | - | 1 | 4435 |