Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK5RAP2 SIRNA Assays ›
Gene Symbol:
CDK5RAP2
Gene Name:
CDK5 regulatory subunit associated protein 2
Gene Aliases:
C48, Cep215, MCPH3
Chromosome Location:
Chr.9: 120388869 - 120580170 on Build GRCh38
UniGene ID:
Hs.269560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26757
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK5RAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011649.2
NP_001011649.1
35
5519
NM_001272039.1
NP_001258968.1
33
5066
NM_018249.5
NP_060719.4
36
5756
NR_073554.1
36
5831
NR_073555.1
36
5754
NR_073556.1
37
5963
NR_073557.1
36
5836
NR_073558.1
36
5833
XM_006717182.1
XP_006717245.1
35
5649
XM_006717185.1
XP_006717248.1
33
5058
XM_011518860.1
XP_011517162.1
36
5742
XM_017014921.1
XP_016870410.1
35
5646
XM_017014922.1
XP_016870411.1
30
4974
XM_017014923.1
XP_016870412.1
32
4821
XM_017014924.1
XP_016870413.1
26
4493
GenBank mRNA
AB046853.2
-
35
5488
AF087969.1
-
3
468
AF448860.1
-
36
5706
AK001729.1
-
14
2594
AK025867.1
-
8
1086
AK027636.1
-
15
2720
AK122913.1
-
36
5745
AL133161.1
-
20
3746
AL833481.1
-
36
5744
BC004526.2
-
9
1365
BC136275.1
-
36
5588
9020262
BC143762.1
-
35
5354
9052276
BC146782.1
-
35
5488
9007237
BQ428975.1
-
1
41
BX537421.1
-
36
5741
BX537759.1
-
35
5506
CR936622.1
-
29
4886
CR936717.1
-
29
4886
KP294527.1
-
36
5756
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
establishment of mitotic spindle orientation
microtubule cytoskeleton organization
microtubule bundle formation
chromosome segregation
centriole replication
brain development
neurogenesis
microtubule organizing center organization
regulation of neuron differentiation
negative regulation of neuron differentiation
positive regulation of transcription, DNA-templated
negative regulation of centriole replication
centrosome organization
regulation of spindle checkpoint
protein binding
calmodulin binding
microtubule binding
tubulin binding
protein kinase binding
protein complex binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0