Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107986910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014152.1 | XP_016869641.1 | 3 | 770 |
Gene Symbol: LOC101929400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014148.1 | XP_016869637.1 | 3 | 1340 |
Gene Symbol: LOC729732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030211.1 | - | 3 | 1028 |