Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD34C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146341.1 | NP_001139813.1 | 2 | 5135 | ||
GenBank mRNA | AK090665.1 | - | 1 | 1519 | ||
AK127037.1 | - | 3 | 4820 | |||
AL109678.1 | - | 2 | 1119 |