Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099851.3 | NP_001093321.1 | 2 | 766 | ||
GenBank mRNA | BC144662.1 | - | 2 | 766 | 9053193 | |
BC146858.1 | - | 2 | 766 | 9056786 |