Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018326.2 | NP_060796.1 | 3 | 1002 | ||
XM_005250017.1 | XP_005250074.1 | 3 | 1018 | |||
GenBank mRNA | AF117333.1 | - | 3 | 970 | ||
AK001972.1 | - | 3 | 964 | |||
AK301441.1 | - | 3 | 1021 | |||
AK301606.1 | - | 4 | 770 | |||
AW836736.1 | - | 145 | ||||
BC020657.1 | - | 3 | 979 | 4701647 |