Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320522.1 | NP_001307451.1 | 11 | 2402 | ||
NM_001320525.1 | NP_001307454.1 | 13 | 2556 | |||
NM_001320526.1 | NP_001307455.1 | 12 | 2621 | |||
NM_001320527.1 | NP_001307456.1 | 11 | 2511 | |||
NM_018332.4 | NP_060802.1 | 12 | 2495 | |||
NR_135296.1 | 10 | 2277 | ||||
GenBank mRNA | AB209401.1 | - | 5 | 4516 | ||
AK094061.1 | - | 2 | 2080 | |||
AL832901.1 | - | 8 | 1979 | |||
AL832970.1 | - | 12 | 2406 | |||
BC005162.2 | - | 12 | 2403 | 3530064 | ||
BC006544.2 | - | 12 | 2403 | 2964377 | ||
CR749227.1 | - | 12 | 2569 |