Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOTAIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003716.3 | 6 | 1675 | |||
NR_047517.1 | 6 | 1681 | ||||
NR_047518.1 | 4 | 1650 | ||||
GenBank mRNA | AK123741.1 | - | 5 | 1487 | ||
DQ926657.1 | - | 6 | 1598 |