Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018352.2 | NP_060822.2 | 4, 5 | 528 | ||
GenBank mRNA | AK002046.1 | - | 4, 5 | 359 | ||
AK298235.1 | - | 4, 5 | 414 | |||
BC011842.2 | - | 4, 5 | 345 | 4110177 | ||
BU159489.1 | - | 4, 5 | 528 |