Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145668.1 | NP_001139140.1 | 2 | 280 | ||
XM_005254378.3 | XP_005254435.1 | 2 | 179 | |||
XM_006720504.2 | XP_006720567.1 | 2 | 162 | |||
XM_011521553.1 | XP_011519855.1 | 2 | 350 | |||
XM_017022178.1 | XP_016877667.1 | 3 | 292 | |||
GenBank mRNA | BX537772.1 | - | 2 | 280 |