Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077620.2 | NP_001071088.1 | 3 | 265 | ||
NR_033357.1 | 3 | 336 | ||||
XM_017025013.1 | XP_016880502.1 | 3 | 2283 | |||
XM_017025014.1 | XP_016880503.1 | 3 | 2283 | |||
XM_017025015.1 | XP_016880504.1 | 3 | 2283 | |||
GenBank mRNA | AK125617.1 | - | 3 | 336 | ||
BC043413.1 | - | 3 | 133 | |||
BC146898.1 | - | 3 | 206 | 9056826 | ||
BC146904.1 | - | 3 | 206 | 9056832 | ||
DQ390338.1 | - | 3 | 272 |