Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HJURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282962.1 | NP_001269891.1 | 6 | 799 | ||
NM_001282963.1 | NP_001269892.1 | 5 | 706 | |||
NM_018410.4 | NP_060880.3 | 8 | 961 | |||
XM_011511437.1 | XP_011509739.1 | 7 | 865 | |||
GenBank mRNA | AB101211.1 | - | 8 | 962 | ||
AB162218.1 | - | 8 | 895 | |||
AK074809.1 | - | 8 | 927 | |||
AK301643.1 | - | 5 | 675 | |||
AK303109.1 | - | 6 | 740 | |||
AL162048.1 | - | 1 | 138 | |||
BC001940.1 | - | 8 | 895 | 2820741 | ||
BM475244.1 | - | 1 | 219 | |||
DN998761.1 | - | 3 | 513 |