Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136536.4 | NP_001130008.2 | 4, 5 | 1163 | ||
NM_001170700.2 | NP_001164171.1 | 4, 5 | 1318 | |||
XM_006714014.3 | XP_006714077.1 | 5, 6 | 1797 | |||
GenBank mRNA | AK131492.1 | - | 4, 5 | 1156 | ||
AK304357.1 | - | 4, 5 | 1149 | |||
AK316523.1 | - | 4, 5 | 1149 | |||
BC157885.1 | - | 4, 5 | 1318 | 9057096 |